Learn More
Fisherbrand™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Brand:  Fisherbrand™
Additional Details : Weight : 10.00000kg
 View more versions of this product
Product Code. 12709165
  • € 27.90 / Pack of 12
Estimated Shipment date
from Supplier 30-07-2020
Add to basket
Description
Description
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Specifications
100 Vials