Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Shop All Thermo Scientific Products
Click to view available options
Holds:
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
Unit Size:
Each
Pack of 12
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
- Use with fiberboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Specifications
| Quantity | 12 Pack |
| Holds | 81 (9 x 9 Grid) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion