missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrillarin Antibody, DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05533C
This item is not returnable.
View return policy
Description
Fibrillarin Monoclonal antibody specifically detects Fibrillarin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| Fibrillarin | |
| Monoclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| EC 2.1.1,34-kD nucleolar scleroderma antigen, EC 2.1.1.-, EC 2.1.1.37, FIB, FIB1,34 kDa nucleolar scleroderma antigen, fibrillarin, FLRNrRNA 2'-O-methyltransferase fibrillarin, RNA, U3 small nucleolar interacting protein 1, RNU3IP1 | |
| Recombinant full length human Fibrillarin sequence expressed in and purified from E. coli. | |
| 0.1 mL | |
| Cellular Markers, Core ESC Like Genes, Neuroscience, Nuclear Envelope Markers, Stem Cell Markers | |
| 2091 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction