Learn More
Invitrogen™ Fibrillarin Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5143506
 View more versions of this product
Product Code. 17836667
  • € 1736.00 / 500µL
Description
Fibrillarin Polyclonal Antibody for Western Blot, ICC/IF, IHC, IP
Nop1p / Fibrillarin was originally identified as a nucleolar protein of bakers yeast, Saccharomyces cerevisiae (accession P15646). The Nop1p protein is essential for yeast viability and is localized in the nucleoli. The human homologue of Nop1p is fibrillarin (accession P22087) a component of the nucleolar small nuclear ribonucleoprotein (snRNP) particle. The human fibrillarin gene is located on chromosome 19 (19q13. 1).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibrillarin
Polyclonal
Unconjugated
fbl
34 kDa nucleolar scleroderma antigen; 34-kD nucleolar scleroderma antigen; AL022665; AT20009p; CELE_T01C3.7; CG5725; CG5725-PA; CG5725-PB; CG5725-PC; CG5725-PD; CG5725-PE; CG5725-PF; CG5725-PG; Dmel\CG5725; Dmel_CG5725; dPANK; dPANK/Fbl; Fbl; fbl-PA; fbl-PB; fbl-PC; fbl-PD; fbl-PE; fbl-PF; fbl-PG; FIB; FIB1; fib-1; fibrillarin; FLRN; fumble; histone-glutamine methyltransferase; LD24618p; LP11571p; nucleolar protein 1; PANK; pantothenate kinase; RNA, U3 small nucleolar interacting protein 1; RNU3IP1; rRNA 2'-O-methyltransferase fibrillarin; T01C3.7
Rabbit
Affinity chromatography
RUO
100064571, 14113, 2091, 292747, 476460
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 5mM sodium azide
P22087, P22509, P35550
fbl
Full length human fibrillarin expressed in and purified from E.Coli.
500 μL
Primary
Canine, Horse, Human, Mouse, Rat