Learn More
Fibrillin 1 Antibody (CL0225) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52885MFV450
Product Code. 30295751
  • € 759.00 / 0.10mL
Description
Fibrillin 1 Monoclonal antibody specifically detects Fibrillin 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibrillin 1
Monoclonal
mFluor Violet 450 SE
Asprosin, FBN, fibrillin 1, fibrillin 1 (Marfan syndrome), fibrillin 15, fibrillin-1, MASS, MFS1, OCTD, SGS, SSKS, WMS
This antibody was developed using a recombinant protein derived from P35555, with the exact immunogen sequence remaining proprietary.
0.1 mL
Extracellular Matrix
2200
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL0225
50mM Sodium Borate
Mouse
Protein A purified