missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrillin 1 Polyclonal Antibody

Product Code. 16399554
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16399554 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16399554 Supplier Invitrogen™ Supplier No. PA582598

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: HCVSGMGMGR GNPEPPVSGE MDDNSLSPEA CYECKINGYP KRGRKRRSTN ETDASNIEDQ SETEANVSLA SWDVEKTAIF AFNISHVSNK VRILELLPAL TTLTNHNRYL IESGNEDGFF KINQKEGISY LHFTKKKPVA GTYSLQISS Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 91%.

Fibrillin 1 is an extracellular acidic protein with a high cysteine content and an extended thread-like shape with mosaic composition of different types of extracellular modules. Most of the fibrillin molecule is contributed by 47 epidermal growth factor-like (EGF-like) repeats; 43 of them have a consensus sequence for calcium binding (cb). Other modules including 8-cysteine motifs (seven), hybrid motifs (two), and unique domains (three) are interspersed throughout the molecule[3-7]. Fibrillin 1 contains a proline-rich region close to the N-terminus that is replaced by a glycine-rich region in Fibrillin 2. The fibrillins are found throughout the connective tissue as integral components of extended fibrils.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillin 1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FBN1
Gene Accession No. P35555
Gene Alias ACMICD; AI536462; Asprosin; B430209H23; ECTOL1; FBN; Fbn1; Fbn-1; Fib-1; fibrillin 1; fibrillin 15; fibrillin-1; GPHYSD2; MASS; MFLS; MFS1; MP340; OCTD; putative; SGS; SSKS; tight skin; Tsk; WMS; WMS2
Gene Symbols FBN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FBN1. Recombinant protein control fragment (Product #RP-90823).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2200
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.