missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrillin 2 Monoclonal Antibody (1C2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524394
This item is not returnable.
View return policy
Description
Peptide Sequence: RQKRSIHEPD PTAVEQISLE SVDMDSPVNM KFNLSHLGSK EHILELRPAI QPLNNHIRYV ISQGNDDSVF RIHQRNGLSY LHTAKKKLMP GTYTLEITSI P.
The protein encoded by this gene is a component of connective tissue microfibrils and may be involved in elastic fiber assembly. Mutations in this gene cause congenital contractural arachnodactyly.
Specifications
| Fibrillin 2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P35556 | |
| FBN2 | |
| FBN2 (NP_001990.2, 2776 a.a. ~ 2876 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1C2 | |
| Unconjugated | |
| FBN2 | |
| BC063774; CCA; DA9; EOMD; Fbn2; Fbn-2; Fib-2; fibrillin 2; fibrillin 5; fibrillin-2; Fibrillin-2 C-terminal peptide; mKIAA4226; sne; sy; syndatyly ems | |
| Mouse | |
| Protein A | |
| RUO | |
| 2201 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction