Learn More
Fibrin Antibody (UC45), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50407
 View more versions of this product
Product Code. 18653818
  • € 372.00 / 0.10mL
Description
Fibrin Monoclonal antibody specifically detects Fibrin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibrin
Monoclonal
1 mg/mL
Flow Cytometry 1:10 - 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready
Mouse
Thiophilic Chromatography
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgM
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA), CyTOF
UC45
Unconjugated
PBS
Human acute monoblastic leukemia cells.
0.1 mL
Apoptosis, Cancer, Cell Biology