missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrin beta-chain Antibody (8E5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-81110PCP
This item is not returnable.
View return policy
Description
Fibrin beta-chain Monoclonal antibody specifically detects Fibrin beta-chain in Human samples. It is validated for ELISA
Specifications
| Fibrin beta-chain | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 8E5 | |
| ELISA, In vivo assay | |
| Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p | |
| 8E5 was prepared fusing Sp2/o myeloma cells with spleen cells derived from female BALB/c mice immunized with a synthetic beta-fibrin peptide seqeunce GHRPLDKC conjugated to MB-KLH (maleimidobenzoylated keyhole limpet hemocyanin). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 2244 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction