Learn More
Fibrinogen Antibody (1F3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB120-10066F
Product Code. 18503173
  • € 470.00 / 0.20mL
Description
Fibrinogen Monoclonal antibody specifically detects Fibrinogen in Human samples. It is validated for Western Blot, Immunoassay, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibrinogen
Monoclonal
FITC
Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with fibrin degradation products.
0.2 mL
Angiogenesis, Cancer, Cell Cycle and Replication
2243
Store at 4°C in the dark.
IgG2b
Western Blot, Immunoassay, Sandwich ELISA
1F3
PBS
Mouse
Protein A purified