missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen Antibody (40F11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 185.00
Specifications
| Antigen | Fibrinogen |
|---|---|
| Clone | 40F11 |
| Dilution | Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Fibrinogen Monoclonal specifically detects Fibrinogen in Human samples. It is validated for Western Blot, Immunoassay, Sandwich ELISA.Specifications
| Fibrinogen | |
| Western Blot 1:100-1:2000, Sandwich ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 2243 | |
| Fibrin degradation products. | |
| Primary | |
| Store at 4°C. Do not freeze. |
| 40F11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| Fib2, fibrinogen alpha chain, fibrinogen, A alpha polypeptide, MGC119422, MGC119423, MGC119425 | |
| FGA | |
| IgG1 | |
| Protein A purified | |
| Fibrinogen, Fibrin degradation products. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title