missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen Beta Chain Monoclonal Antibody (2#)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA541586

 View more versions of this product

Product Code. 17857102

  • € 476.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Fibrinogen Beta Chain Monoclonal Antibody for Western Blot, IHC (P)

FIBB is the beta component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in its gene lead to several disorders, including afibrinogenemia, dysfibrinogenemia, hypodysfibrinogenemia and thrombotic tendency.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fibrinogen Beta Chain
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P02675
Fgb
Recombinant protein Fibrinogen Beta Chain. The antigen corresponds to amino acid range 45-491 of the target protein.
200 μL
Primary
Human
Antibody
IgG2b κ
Immunohistochemistry (Paraffin), Western Blot
2#
Unconjugated
Fgb
2510049G14Rik; Ab1-181; Ab1-216; Ac1-581; B beta polypeptide; beta-fibrinogen; epididymis secretory sperm binding protein Li 78p; Fgb; fibrinogen; Fibrinogen beta chain; fibrinogen, B beta polypeptide; fibrinogen, beta polypeptide; Fibrinopeptide B; HEL-S-78p; Liver regeneration-related protein LRRG036/LRRG043/LRRG189; MGC104327; MGC120405
Mouse
Protein A/G
RUO
2244
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.