missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen gamma chain Antibody (5A6), Novus Biologicals™
Mouse Monoclonal Antibody
€ 210.00 - € 533.00
Specifications
| Antigen | Fibrinogen gamma chain |
|---|---|
| Clone | 5A6 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18104418
|
Novus Biologicals
NBP2-37560 |
0.1 mL |
€ 533.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648705
|
Novus Biologicals
NBP2-37560-0.025ml |
0.025 mL |
€ 210.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibrinogen gamma chain Monoclonal specifically detects Fibrinogen gamma chain in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Fibrinogen gamma chain | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| fibrinogen gamma chain, fibrinogen, gamma polypeptide | |
| FGG | |
| IgG2b | |
| Unpurified |
| 5A6 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer | |
| Ascites with 0.03% Sodium Azide | |
| 2266 | |
| Purified recombinant fragment of human FGG expressed in E. Coli. | |
| Primary | |
| 52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title