missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fibrinogen gamma Monoclonal Antibody (546CT11.4.3)
GREENER_CHOICE

Product Code. 17167188 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167188 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17167188 Supplier Invitrogen™ Supplier No. MA537602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fibrinogen gamma Monoclonal Antibody for Western Blot

The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrinogen gamma
Applications Western Blot
Classification Monoclonal
Clone 546CT11.4.3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.09% sodium azide; pH 7.4
Gene Fgg
Gene Accession No. P02679
Gene Alias 3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951)
Gene Symbols Fgg
Host Species Mouse
Immunogen KLH conjugated synthetic peptide between 417-445 amino acids from human FGG.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2266
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.