missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibrinogen gamma Monoclonal Antibody (K24024_2E1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555847
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: KVDKDLQSLE DILHQVENKT SEVKQLIKAI QLTYNPDESS KPNMIDAATL KSRKMLEEIM KYEASILTHD SSIRYLQEIY NSNNQKIVNL KEKVAQLEAQ
The protein encoded by this gene is the gamma component of fibrinogen, a blood-borne glycoprotein comprised of three pairs of nonidentical polypeptide chains. Following vascular injury, fibrinogen is cleaved by thrombin to form fibrin which is the most abundant component of blood clots. In addition, various cleavage products of fibrinogen and fibrin regulate cell adhesion and spreading, display vasoconstrictor and chemotactic activities, and are mitogens for several cell types. Mutations in this gene lead to several disorders, including dysfibrinogenemia, hypofibrinogenemia and thrombophilia. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications
| Fibrinogen gamma | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P02679 | |
| Fgg | |
| Recombinant human FGG (immunogen range is 61-160). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| K24024_2E1 | |
| Unconjugated | |
| Fgg | |
| 3010002H13Rik; AI256424; FGG; fibg; fibrinogen; fibrinogen gamma chain; Fibrinogen gamma-B chain; fibrinogen gamma-chain; fibrinogen, gamma polypeptide; gamma'; gamma-fibrinogen; PRO2061; testicular tissue protein Li 70; Unknown (protein for MGC:127951) | |
| Mouse | |
| Protein A | |
| RUO | |
| 2266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction