Learn More
Fibroblast Activation Protein alpha/FAP Antibody (427819) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB3715JF525
Product Code. 30175176
  • ONLINE EXCLUSIVE PRICE
    € 661.00
    € 624.75 / 0.10mL
    Save € 36.25
    5% Off
Description
Fibroblast Activation Protein alpha/FAP Monoclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Human samples. It is validated for Flow Cytometry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibroblast Activation Protein alpha/FAP
Monoclonal
Janelia Fluor 525
170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Fibroblast Activation Protein alpha /FAP, Leu26-Asp760, Accession # Q12884
0.1 mL
Cancer, Protein Phosphatase
2191
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF
427819
50mM Sodium Borate
Mouse
Protein A or G purified