missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Activation Protein alpha/FAP Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-85534PCP
This item is not returnable.
View return policy
Description
Fibroblast Activation Protein alpha/FAP Polyclonal antibody specifically detects Fibroblast Activation Protein alpha/FAP in Mouse samples. It is validated for Western Blot, ELISA
Specifications
| Fibroblast Activation Protein alpha/FAP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, ELISA | |
| PerCP | |
| 170 kDa melanoma membrane-bound gelatinase, DKFZp686G13158, DPPIV, EC 3.4.21.-, FAPA, Fibroblast activation protein alpha, fibroblast activation protein, alpha, Integral membrane serine protease, seprase, vibronectin | |
| This affinity purified Fibroblast Activation Protein alpha/FAP Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of mouse Fibroblast Activation Protein alpha/Fibroblast Activation Protein alpha/FAP protein. (Uniprot: P97321) | |
| 0.1 mL | |
| Cancer, Protein Phosphatase | |
| 2191 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction