missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibroblast Antibody (ER-TR7), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64932C
This item is not returnable.
View return policy
Description
Fibroblast Monoclonal specifically detects Fibroblast in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifikationer
| Fibroblast | |
| Monoclonal | |
| DyLight 650 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Rat | |
| 0.1 mL | |
| NB100-64932 recognizes ER-TR7, an antigen that is located in the cytoplasm of reticular fibroblasts and is a component of the extracellular matrix of lymphoid and non-lymphoid organs. The antigen recognized by clone ER-TR7 has not been identified but studies suggest that it is likely to be distinct from laminin, fibronectin, collagen types I-IV, heparin sulphate proteoglycan, entactin and nidogen. Clone ER-TR7 has been used to stain the microanatomy of various organs and also stains subendothelial deposits in atherosclerotic plaques. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| ER-TR7 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| reticular fibroblast | |
| Isolated C3H thymic stromal cells | |
| Primary | |
| Mouse | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering