Learn More
Fibronectin Antibody (1E1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26577-50ul
 View more versions of this product
Product Code. 30043724
  • € 355.00 / 50µL
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibronectin
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
1E1
ELISA, Immunohistochemistry 1:50-1:200
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
A synthesized peptide derived from Human Fibronectin [UniProt P02751]
50 μL
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
2335
Store at -20 to -70C. Avoid freeze-thaw cycles.