missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (960632), Alexa Fluor™ 488, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB8258AF488
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal specifically detects Fibronectin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Fibronectin | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FN1 | |
| Partial recombinant human Fibronectin/Anastellin protein (amino acids 631-705) [UniProt P02751] | |
| 100 μL | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Neuronal Stem Cell Markers, Stem Cell Markers | |
| 2335 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 960632 | |
| Western Blot, Immunohistochemistry | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction