Learn More
Fibronectin Antibody (A17), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-23524FR
Product Code. 18494406
  • ONLINE EXCLUSIVE PRICE
    € 628.00
    € 593.25 / 100µL
    Save € 34.75
    5% Off
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human, Bovine, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibronectin
Monoclonal
DyLight 680
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Lysed bovine corneal endothelial cells and extracellular matrix
100 μg
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
2335
Store at 4°C in the dark.
IgG1
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
A17
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Bovine, Chicken