Learn More
Fibronectin Antibody (CL3730) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61633F
Product Code. 30299960
  • € 791.00 / 0.10mL
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibronectin
Monoclonal
FITC
CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
This antibody was developed using a recombinant protein derived from P02751, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
2335
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL3730
PBS
Mouse
Protein A purified