missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 1 Antibody (CL0337), Novus Biologicals™
Mouse Monoclonal Antibody
€ 369.00 - € 564.00
Specifications
| Antigen | Fibulin 1 |
|---|---|
| Clone | CL0337 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699648
|
Novus Biologicals
NBP2-52918-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670479
|
Novus Biologicals
NBP2-52918 |
0.1 mL |
€ 564.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibulin 1 Monoclonal antibody specifically detects Fibulin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| Fibulin 1 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| FBLN, FIBL1, FIBL-1, fibulin 1, fibulin-1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: CESGIHNCLPDFICQNTLGSFRCRPKLQCKSGFIQDALGNCIDINECLSISAPCPIGHTCINTEGSYTCQKNVPNCGRGYHLNEEGTRCVDVDECAPPAEPCGKGHRCVNSPGSFRCECKTGYYFDGISRMCVDVNE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL0337 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS (pH 7.2), 40% Glycerol | |
| 2192 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title