Learn More
Fibulin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33479
This item is not returnable.
View return policy
Description
Fibulin 2 Polyclonal specifically detects Fibulin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Fibulin 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| FBLN2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human Fibulin 2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 2199 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| FIBL-2, fibulin 2, fibulin-2 | |
| Rabbit | |
| 127 kDa | |
| 100 μL | |
| Primary | |
| Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |