missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin-3/EFEMP1 Antibody (OTI1D9) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-70700
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Fibulin-3/EFEMP1 Monoclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Spezifikation
| Fibulin-3/EFEMP1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:200-500, Flow Cytometry 1:100, CyTOF-ready | |
| DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5 | |
| Mouse | |
| 52.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, CyTOF | |
| OTI1D9 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| EFEMP1 | |
| Full length human recombinant protein of human EFEMP1(NP_004096) produced in HEK293T cell. | |
| Affinity Purified | |
| Vision | |
| 2202 | |
| Human, Mouse, Rat | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur