missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibulin 3 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
€ 208.00 - € 504.00
Specifications
| Antigen | Fibulin 3 |
|---|---|
| Clone | C1 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17847102
|
Invitrogen™
MA541587 |
200 μL |
€ 504.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17644276
|
Invitrogen™
MA543820 |
20 μL |
€ 208.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fibulin 3 Monoclonal Antibody for Western Blot
This gene encodes a member of the fibulin family of extracellular matrix glycoproteins. Like all members of this family, the encoded protein contains tandemly repeated epidermal growth factor-like repeats followed by a C-terminus fibulin-type domain. This gene is upregulated in malignant gliomas and may play a role in the aggressive nature of these tumors. Mutations in this gene are associated with Doyne honeycomb retinal dystrophy. Alternatively spliced transcript variants that encode the same protein have been described.Specifications
| Fibulin 3 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O35568, Q12805 | |
| 2202, 305604 | |
| Recombinant protein Fibulin 3. The antigen corresponds to amino acid range 26-213 of the target protein. | |
| Antibody |
| C1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human, Rat | |
| EFEMP1 | |
| DHRD; DRAD; EFEMP1; EGF containing fibulin like extracellular matrix protein 1; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; epidermal growth factor-containing fibulin-like extracellular matrix protein 1; extracellular protein S1-5; FBLN3; FBNL; FIBL-3; Fibrillin-like protein; Fibulin-3; FLJ35535; MGC111353; MLVT; MTLV; S1-5; T16 | |
| EFEMP1 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title