Learn More
Invitrogen™ Fibulin 3 Recombinant Rabbit Monoclonal Antibody (HL2572)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556691
Product Code. 30276171
  • € 500.00 / 100µL
Description
Fibulin 3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the fibulin family of extracellular matrix glycoproteins. Like all members of this family, the encoded protein contains tandemly repeated epidermal growth factor-like repeats followed by a C-terminus fibulin-type domain. This gene is upregulated in malignant gliomas and may play a role in the aggressive nature of these tumors. Mutations in this gene are associated with Doyne honeycomb retinal dystrophy. Alternatively spliced transcript variants that encode the same protein have been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fibulin 3
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q12805
EFEMP1
Recombinant fragment of human EFEMP1 (aa 193-483).
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
HL2572
Unconjugated
EFEMP1
DHRD; DRAD; EFEMP1; EGF containing fibulin like extracellular matrix protein 1; EGF containing fibulin-like extracellular matrix protein 1; EGF-containing fibulin-like extracellular matrix protein 1; epidermal growth factor-containing fibulin-like extracellular matrix protein 1; extracellular protein S1-5; FBLN3; FBNL; FIBL-3; Fibrillin-like protein; Fibulin-3; FLJ35535; MGC111353; MLVT; MTLV; S1-5; T16
Rabbit
Protein A
RUO
2202
-20°C, Avoid Freeze/Thaw Cycles