missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibulin 5/DANCE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16495
This item is not returnable.
View return policy
Description
Fibulin 5/DANCE Polyclonal specifically detects Fibulin 5/DANCE in Human samples. It is validated for Western Blot.
Specifications
| Fibulin 5/DANCE | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| ARMD3fibulin-5, Dance, DANCEFIBL-5, Developmental arteries and neural crest EGF-like protein, EVEC, fibulin 5, UP50FLJ90059, Urine p50 protein | |
| Rabbit | |
| 50 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (100%), Rat (100%), Bovine (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9UBX5 | |
| FBLN5 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Fibulin 5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 10516 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction