missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibulin 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557106
This item is not returnable.
View return policy
Description
Immunogen sequence: ASQNCLSKQQ LLSAIRQLQQ LLKGQETRFA EGIRHMKSRL AALQNSVGRV GPDALPVSCP ALNTPADGRK FGSKYLVDHE Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 75%.
An adhesion molecule that interacts with extracellular matrix molecules in developing teeth and may play important roles in differentiation and maintenance of odontoblasts as well as in dentin formation.
Specifications
| Fibulin 7 | |
| Polyclonal | |
| Unconjugated | |
| FBLN7 | |
| 1600015H20Rik; AI464334; extracellular matrix protein TM14; Fbln7; FIBL-7; fibulin 7; fibulin-7; Protein TM14; RGD1310131; TM14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 129804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q53RD9 | |
| FBLN7 | |
| Recombinant protein corresponding to Human Fibulin 7. Recombinant protein control fragment (Product #RP-96117). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction