missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibulin 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532125
This item is not returnable.
View return policy
Description
Recommended positive controls: HCT116, MCF-7. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
An adhesion molecule that interacts with extracellular matrix molecules in developing teeth and may play important roles in differentiation and maintenance of odontoblasts as well as in dentin formation.
Specifications
| Fibulin 7 | |
| Polyclonal | |
| Unconjugated | |
| FBLN7 | |
| 1600015H20Rik; AI464334; extracellular matrix protein TM14; Fbln7; FIBL-7; fibulin 7; fibulin-7; Protein TM14; RGD1310131; TM14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 129804 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.88 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q53RD9 | |
| FBLN7 | |
| Recombinant protein encompassing a sequence within the center region of human Fibulin 7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction