Learn More
FICD Monoclonal Antibody (OTI3E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802903
Encompass_Preferred
 View more versions of this product
Product Code. 17551713
  • € 540.87 / 100µL
Description
The function remains known.
Specifications
Specifications
FICD
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BVA6
FICD
Human recombit protein fragment corresponding to amino acids 61-265 of human FICD produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3E3
Unconjugated
FICD
HIP13, HYPE, UNQ3041
Mouse
Affinity Chromatography
RUO
11153
-20° C, Avoid Freeze/Thaw Cycles