Learn More
FICD Monoclonal Antibody (OTI3E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802903S
Encompass_Preferred
 View more versions of this product
Product Code. 17571713
  • € 242.00 / 30µL
Description
FICD Monoclonal antibody specifically detects FICD in Human samples. It is validated for Western Blot
Specifications
Specifications
FICD
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BVA6
Mouse
Affinity chromatography
RUO
11153
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3E3
Unconjugated
Ficd
adenosine monophosphate-protein transferase FICD; AMPylator FICD; D5Ertd40e; De-AMPylase FICD; FIC domain containing; FIC domain protein adenylyltransferase; FIC domain-containing protein; fic S-phase protein cell division homolog; FICD; HIP13; HIP-13; huntingtin interacting protein 13; huntingtin interacting protein E; huntingtin interactor protein E; huntingtin yeast partner E; Huntingtin-interacting protein 13; huntingtin-interacting protein E; Hype; MGC5623; Protein adenylyltransferase FICD; protein adenylyltransferase FICD; adenosine monophosphate-protein transferase FICD; UNQ3041; UNQ3041/PRO9857
Human recombinant protein fragment corresponding to amino acids 61-265 of human FICD produced in E.coli.
30 μL