Learn More
Ficolin-1 Mouse anti-Human, Clone: 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05050
This item is not returnable.
View return policy
Description
Ficolin-1 Monoclonal specifically detects Ficolin-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ficolin-1 | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1:20000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Collagen/fibrinogen domain-containing protein 1, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin-A, Ficolin-alpha, M-ficolin | |
| Purified prokariot expressed recombinant human M-Ficolin (aa 95 to 314) | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2219 | |
| Store at 4°C. Do not freeze. | |
| IgG1 κ |