Learn More
Ficolin-1 Mouse anti-Human, Clone: 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP1-05050
 View more versions of this product
Product Code. 18555402
  • € 522.00 / 0.10mg
Description
Ficolin-1 Monoclonal specifically detects Ficolin-1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ficolin-1
Monoclonal
1 mg/mL
ELISA 1:20000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Collagen/fibrinogen domain-containing protein 1, FCNMficolin-A, ficolin (collagen/fibrinogen domain containing) 1, ficolin (collagen/fibrinogen domain-containing) 1, ficolin-1, Ficolin-A, Ficolin-alpha, M-ficolin
Purified prokariot expressed recombinant human M-Ficolin (aa 95 to 314)
0.1 mg
Primary
Human
Purified
ELISA
1
Unconjugated
10mM Phosphate (pH 7.4) and 0.5M NaCl
Mouse
Protein A or G purified
RUO
2219
Store at 4°C. Do not freeze.
IgG1 κ