missing translation for 'onlineSavingsMsg'
Learn More

Fidaxomicin, MedChemExpress

Product Code. 16413437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413437 50 mg 50mg
16489877 5 mg 5mg
16437408 10 mg 10mg
16407386 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16413437 Supplier Medchem Express Supplier No. HY17580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fidaxomicin (OPT-80), a macrocyclic antibiotic, is an orally active and potent RNA polymerase inhibitor. Fidaxomicin has a narrow spectrum of antibacterial activity and a good anti-Clostridium difficile activity (MIC90=0.12 μg/mL). Fidaxomicin can be used for Clostridium difficile infection (CDI) research.

Fidaxomicin (OPT-80), a macrocyclic antibiotic, is an orally active and potent RNA polymerase inhibitor. Fidaxomicin has a narrow spectrum of antibacterial activity and a good anti-Clostridium difficile activity (MIC90=0.12 μg/mL). Fidaxomicin can be used for Clostridium difficile infection (CDI) research.

Specifications

Chemical Name or Material Fidaxomicin
CAS 873857-62-6
Color White
Purity Grade Notes Research
Molecular Formula C52H74Cl2O18
Quantity 50 mg
Synonym OPT-80 PAR-101
Solubility Information DMSO : ≥ 33 mg/mL (31.19 mM)
SMILES O=C(C(C(O)=C(Cl)C(O)=C1Cl)=C1CC)O[C@H]([C@H](O[C@H]2OC/C(C(O[C@](C/C=C3C)([H])[C@H](O)C)=O)=C\C=C\C[C@@H](/C(C)=C\[C@H](CC)[C@@H](O[C@@](OC(C)(C)[C@@H](OC(C(C)C)=O)[C@@H]4O)([H])[C@H]4O)\C(C)=C/3)O)C)[C@@H]([C@@H]2OC)O
Molecular Weight (g/mol) 1058.04
Formula Weight 1058.04
Percent Purity 96.8%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.