missing translation for 'onlineSavingsMsg'
Learn More

Fidaxomicin, MedChemExpress

Product Code. 16489877
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16489877

Brand: Medchem Express HY17580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fidaxomicin (OPT-80), a macrocyclic antibiotic, is an orally active and potent RNA polymerase inhibitor. Fidaxomicin has a narrow spectrum of antibacterial activity and a good anti-Clostridium difficile activity (MIC90=0.12 μg/mL). Fidaxomicin can be used for Clostridium difficile infection (CDI) research.

Fidaxomicin (OPT-80), a macrocyclic antibiotic, is an orally active and potent RNA polymerase inhibitor. Fidaxomicin has a narrow spectrum of antibacterial activity and a good anti-Clostridium difficile activity (MIC90=0.12 μg/mL). Fidaxomicin can be used for Clostridium difficile infection (CDI) research.

Specifications

Chemical Name or Material Fidaxomicin
CAS 873857-62-6
Color White
Purity Grade Notes Research
Molecular Formula C52H74Cl2O18
Quantity 5 mg
Synonym OPT-80 PAR-101
Solubility Information DMSO : ≥ 33 mg/mL (31.19 mM)
SMILES O=C(C(C(O)=C(Cl)C(O)=C1Cl)=C1CC)O[C@H]([C@H](O[C@H]2OC/C(C(O[C@](C/C=C3C)([H])[C@H](O)C)=O)=C\C=C\C[C@@H](/C(C)=C\[C@H](CC)[C@@H](O[C@@](OC(C)(C)[C@@H](OC(C(C)C)=O)[C@@H]4O)([H])[C@H]4O)\C(C)=C/3)O)C)[C@@H]([C@@H]2OC)O
Molecular Weight (g/mol) 1058.04
Formula Weight 1058.04
Percent Purity 96.8%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.