Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Brand: Thermo Scientific™ FLN4567LE
1855.00 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 15.00000kg
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:- LN2 backup system maintains temperature down to -80°C with liquid nitrogen
- CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
Specifications
LN2 Backup System: maintains temperature down to -80°C with liquid nitrogen | |
-80°C | |
Boxes (300) |
Field-installed | |
Thermo Scientific TSX Series |