missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIGLA Antibody (6A10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00344018-M03-100ug
This item is not returnable.
View return policy
Description
FIGLA Monoclonal antibody specifically detects FIGLA in Human samples. It is validated for Western Blot, ELISA
Specifications
| FIGLA | |
| Monoclonal | |
| Unconjugated | |
| NP_001004311.1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 344018 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 6A10 | |
| In 1x PBS, pH 7.4 | |
| BHLHC8, Class C basic helix-loop-helix protein 8, factor in the germline alpha, FIGalpha, folliculogenesis specific basic helix-loop-helix, Folliculogenesis-specific basic helix-loop-helix protein, POF6, Transcription factor FIGa | |
| FIGLA (NP_001004311.1, 110 a.a. ∽ 219 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ATEYIQVLSDLLEGAKDSKKQDPDEQSYSNNTSESHTSSARQLSRNITQHISCAFGLKNEEEGPWADGGSGEPAHACRHSVMSTTEIISPTRSLDRFPEVELLSHRLPQV | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction