missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIGLA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-66846
This item is not returnable.
View return policy
Description
FIGLA Polyclonal specifically detects FIGLA in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| FIGLA | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide | |
| FIGLA | |
| Synthetic peptide within Human FIGLA aa 21-70/ 219. (SwissProt: O55208 Mouse) | |
| 100 μL | |
| 344018 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:100 | |
| BHLHC8, Class C basic helix-loop-helix protein 8, factor in the germline alpha, FIGalpha, folliculogenesis specific basic helix-loop-helix, Folliculogenesis-specific basic helix-loop-helix protein, POF6, Transcription factor FIGa | |
| Rabbit | |
| Immunogen affinity purified | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction