missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIGNL1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2355H
This item is not returnable.
View return policy
Description
FIGNL1 Polyclonal antibody specifically detects FIGNL1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| FIGNL1 | |
| Polyclonal | |
| Western Blot | |
| EC 3.6.4, EC 3.6.4.-, fidgetin-like 1, fidgetin-like protein 1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Fidgetin-Like 1using the numbering given in entry NP_071399.2 (GeneID 63979). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 63979 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction