missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIGNL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2355
This item is not returnable.
View return policy
Description
FIGNL1 Polyclonal specifically detects FIGNL1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| FIGNL1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 3.6.4, EC 3.6.4.-, fidgetin-like 1, fidgetin-like protein 1 | |
| Rabbit | |
| 74 kDa | |
| 100 μL | |
| Primary | |
| Human, Mouse. Based on 100% sequence identity, this antibody is predicted to react with Rabbit, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, African elephant, Naked mole rat and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 2-5 ug/mg lysate | |
| Q6PIW4 | |
| FIGNL1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Fidgetin-Like 1using the numbering given in entry NP_071399.2 (GeneID 63979). | |
| Affinity Purified | |
| RUO | |
| 63979 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction