missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIH-1/HIF-1AN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 307.65
Specifications
| Antigen | FIH-1/HIF-1AN |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606878
|
Novus Biologicals
NBP2-99356-100ul |
100 μg |
€ 326.00 € 307.65 / 100µL Save € 18.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622897
|
Novus Biologicals
NBP2-99356-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FIH-1/HIF-1AN Polyclonal antibody specifically detects FIH-1/HIF-1AN in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| FIH-1/HIF-1AN | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Chromatin Research, Hypoxia, Transcription Factors and Regulators | |
| 0.2 um filtered solution in PBS | |
| 55662 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp762F1811, EC 1.14.11.16, factor inhibiting HIF1, Factor inhibiting HIF-1, FIH1FIH-1, FLJ20615, FLJ22027, hypoxia inducible factor 1, alpha subunit inhibitor, hypoxia-inducible factor 1-alpha inhibitor, Hypoxia-inducible factor asparagine hydroxylase, peptide-aspartate beta-dioxygenase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FIH-1/HIF-1AN. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title