missing translation for 'onlineSavingsMsg'
Learn More

Filaggrin (Keratinocyte Differentiation Marker) Mouse, Clone: FLG/1563, enQuireBio™

Mouse Monoclonal Antibody

€ 199.00 - € 478.00

Specifications

Antigen Filaggrin (Keratinocyte Differentiation Marker)
Clone FLG/1563
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15929997
View Documents
EnquireBio
2312-MSM3-P0
20 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15939997
View Documents
EnquireBio
2312-MSM3-P1
100 μg
ONLINE EXCLUSIVE PRICE
€ 554.40
€ 478.00 / 100µg
Save € 76.40
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Filaggrin (Keratinocyte Differentiation Marker)
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant human Filaggrin protein fragment (apprx. aa 998-1104)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
26-45kDa (Processed); 350kDa (Profilaggrin)
FLG/1563
Immunohistochemistry
Unconjugated
RUO
2312
IgG2b κ
Protein A/G purified from Bioreactor concentrate.
Cytoplasmic
U-251-MG cells or Skin
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.