missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Filaggrin Polyclonal Antibody
GREENER_CHOICE

Product Code. 16370085
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16370085 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16370085 Supplier Invitrogen™ Supplier No. PA583128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RKRLSERLEE KEDNEEGVYD YENTGRMTQK WIQSGHIATY YTIQDEAYDT TDSLLEENKI YERSRSSDGK SSSQVNRSRH ENTSQVPLQE SR Highest antigen sequence indentity to the following orthologs: Mouse - 28%, Rat - 27%.

Filaggrin is expressed only in well differentiated keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filaggrin
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FLG
Gene Accession No. P20930
Gene Alias ATOD2; AW107830; epidermal filaggrin; filaggrin; Filaggrin (profilaggrin); FLG; ft; Pflg
Gene Symbols FLG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FLG. Recombinant protein control fragment (Product #RP-94297).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2312
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.