missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Filamin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582234
This item is not returnable.
View return policy
Description
Immunogen sequence: HSLHETSTVL VETVTKSSSS RGSSYSSIPK FSSDASKVVT RGPGLSQAFV GQKNSFTVDC SKAGTNMMMV GVHGPKTPCE EVYVKHMGNR VYNVTYTVKE KGDY Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
This gene encodes one of three related filamin genes, specifically gamma filamin. These filamin proteins crosslink actin filaments into orthogonal networks in cortical cytoplasm and participate in the anchoring of membrane proteins for the actin cytoskeleton. Three functional domains exist in filamin: an N-terminal filamentous actin-binding domain, a C-terminal self-association domain, and a membrane glycoprotein-binding domain. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Filamin 2 | |
| Polyclonal | |
| Unconjugated | |
| FLNC | |
| 1110055E19Rik; ABP-280; ABP280A; ABP-280-like protein; ABPA; ABPL; ABP-L; ABP-L, gamma filamin; actin binding protein 280; actin-binding protein 280; actin-binding-like protein; filamin C; Filamin C gamma; filamin C, gamma; filamin C, gamma (actin binding protein 280); filamin, gamma; filamin-2; Filamin-C; FLJ10186; FLN2; FLNC; FLN-C; gamma filamin; gamma-filamin; MFM5; MPD4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2318 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14315 | |
| FLNC | |
| Recombinant protein corresponding to Human FLNC. Recombinant protein control fragment (Product #RP-88627). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction