missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Filamin B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552129
This item is not returnable.
View return policy
Description
Immunogen sequence: HGKVGEAGLL SVDCSEAGPG ALGLEAVSDS GTKAEVSIQN NKDGTYAVTY VPLTAGMYTL TMKYGGELVP HFPARVKVEP AVDTSRIKVF GPGIEGKDVF REATTDFTV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 89%.
FOXK1 is expressed in these cells and regulates cell cycle progression through an interaction with its downstream target the cyclin-dependent kinase inhibitor p21 (CIP). Loss of FOXK1 in mice results in growth retardation and a severe impairment in skeletal muscle regeneration following injury. FOXK1 also shows expression in immature tissues of brain, eye, heart, lung and thymus. It also is predominantly expressed in many malignant tissues, such as tumors of the brain, colon and lymph node.
Specifications
| Filamin B | |
| Polyclonal | |
| Unconjugated | |
| FLNB | |
| ABP-278; ABP-280; ABP-280 homolog; ABP-280-like protein; actin binding protein 278; actin-binding-like protein; AL024016; AOI; beta-filamin; Fh1; filamin B; filamin B, beta; filamin homolog 1; filamin, beta; filamin-3; FilaminB; Filamin-B; FLN1L; FLN3; FLNB; FLN-B; Larsen syndrome 1 (autosomal dominant); LRS1; SCT; TABP; TAP; thyroid autoantigen; Truncated ABP; Truncated actin-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2317 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75369 | |
| FLNB | |
| Recombinant protein corresponding to Human Filamin B. Recombinant protein control fragment (Product #RP-88633). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction