missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filensin Antibody (OTI2B2), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70249AF647
This item is not returnable.
View return policy
Description
Filensin Monoclonal specifically detects Filensin in Human samples. It is validated for Western Blot.
Specifications
| Filensin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BFSP1 | |
| Recombinant protein fragment corresponding to amino acids 405-665 of human Filensin (NP_001186) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI2B2 | |
| Western Blot | |
| Beaded filament structural protein 1, beaded filament structural protein 1, filensin, CP115Lens intermediate filament-like heavy, filensin, Lens fiber cell beaded-filament structural protein CP 115, LIFL-H115 KD | |
| Mouse | |
| Affinity Purified | |
| Vision | |
| 631.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction