missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Filensin Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA559279

Product Code. 15727141

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SAHECHDDEI QLYNEQIETL RKEIEETERV LEKSSYDCRQ LAVAQQTLKN ELDRYHRIIE IEGNRLTSAF IETPIPLFTQ SHGVSLSTGS GGKD Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 82%.

More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Filensin
Polyclonal
Unconjugated
BFSP1
Beaded filament structural protein 1; beaded filament structural protein 1, filensin; BFSP1; CP115; CP94; CTRCT33; cytoskeletal protein, 115 KD; filensin; Lens fiber cell beaded-filament structural protein CP 115; lens intermediate filament-like heavy; LIFL-H; LOW QUALITY PROTEIN: filensin; RP4-531H16.2
Rabbit
Antigen affinity chromatography
RUO
631
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q12934
BFSP1
Recombinant protein corresponding to Human Filensin. Recombinant protein control fragment (Product #RP-99408).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.