missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FILIP1L Monoclonal Antibody (4H7G5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550279
This item is not returnable.
View return policy
Description
FILIP1L Monoclonal Antibody for Western Blot, ELISA
Acts as a regulator of the antiangiogenic activity on endothelial cells. When overexpressed in endothelial cells, leads to inhibition of cell proliferation and migration and an increase in apoptosis. Inhibits melanoma growth When expressed in tumor-associated vasculature.
Specifications
| FILIP1L | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Q4L180 | |
| FILIP1L | |
| Recombinant Human Filamin A-interacting protein 1-like protein (1-200AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 4H7G5 | |
| Unconjugated | |
| FILIP1L | |
| 130 kDa GPBP-interacting protein; 4631422O05Rik; 90 kDa GPBP-interacting protein; COL4A3BPIP; DOC1; DOC-1; downregulated in ovarian cancer 1-like; filamin A interacting protein 1 like; filamin A interacting protein 1-like; filamin A-interacting protein 1-like; FILIP1L; GIP130; GIP130a; GIP130b; GIP130c; GIP90; Protein down-regulated in ovarian cancer 1; protein down-regulated in ovarian cancer 1 homolog; RGD1565927 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11259 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction