Learn More
3M™ Filter
For effective protection against a range of particulates, gases and vapours or combination hazards, use 3M™ Filters DT-Series.
€ 126.00 - € 344.00
Specifications
Product Type | Filter |
---|---|
Size | Standard |
For Use With (Equipment) | 3M Air Respirators |
Product Code | Brand | Cartridge/Filter | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cartridge/Filter | Price | Quantity & Availability | |||||
17863246
|
3M™
7100260622 |
PARTICULATE |
€ 126.00
Pack of 10 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
17883246
|
3M™
7100260628 |
COMBINATION |
€ 344.00
Pack of 10 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Description
3M™ Filters DT-Series are available in a range of specifications to provide protection against particulate hazards, gas & vapour hazards or a combination of both. They are designed for use with 3M Reusable and Powered Air Respirators with DIN 40 filter ports.
Specifications
Filter | |
Standard | |
3M Air Respirators |