Learn More
Duran™ Filter Candle Without Tube
The Filter Candle Without Tube is suited to applications where the filtration of very small quantities is required. This filtration is possible from a liquid or a carrier gas.
POA
Specifications
Quantity 1 Ea.
Length (Metric) 65 mm
View More Specs
Description
Description
  • Without tube
  • Duran™ borosilicate glass 3.3
  • Can be used as a filter to separate the smallest amount of solids from the filter surface
Specifications
Specifications
1 Ea.
65 mm